missing translation for 'onlineSavingsMsg'
Learn More

1,8-Dibromoanthracene, TRC

Product Code. 30384618
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30384618 50 mg
30374916 100 mg
30376436 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30384618 Supplier Toronto Research Chemicals Supplier No. TRCD42350850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131276-24-9
Molecular Formula C14H8Br2
Molecular Weight (g/mol) 336.02
SMILES Brc1cccc2cc3cccc(Br)c3cc12

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C14H8Br2/c15-13-5-1-3-9-7-10-4-2-6-14(16)12(10)8-11(9)13/h1-8H
Formula Weight 333.8993
Chemical Name or Material 1,8-Dibromoanthracene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.