CAS RN 131276-24-9
IUPAC Name:
1,8-dibromoanthracene
Synonyms:
1,8-dibromoanthracene
Molecular Weight (g/mol):
336.03
Molecular Formula:
C14H8Br2
InChi Key:
JQQFVBWHOMXEEA-UHFFFAOYSA-N
SMILES:
BrC1=CC=CC2=CC3=CC=CC(Br)=C3C=C12
1
–
1
of
1
results
1
1,8-Dibromoanthracene, TRC
CAS: 131276-24-9 Molecular Formula: C14H8Br2 Molecular Weight (g/mol): 336.02 SMILES: Brc1cccc2cc3cccc(Br)c3cc12