missing translation for 'onlineSavingsMsg'
Learn More

[1-(6-Methylpyrazin-2-yl)piperid-4-yl]methylamine, 97%, Thermo Scientific™

Product Code. 10557284
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10557284 1 g Amber Glass Bottle 1g
10316244 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10557284 Supplier Thermo Scientific Maybridge Supplier No. CC58913DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 892502-20-4
Molecular Formula C11H18N4
Molecular Weight (g/mol) 206.293
MDL Number MFCD09025897
InChI Key XVEZCDMRMDBGQU-UHFFFAOYSA-N
Synonym 1-6-methylpyrazin-2-yl piperid-4-yl methylamine, 1-6-methylpyrazin-2-yl piperidin-4-yl methanamine, 1-1-6-methylpyrazin-2-yl piperidin-4-yl methanamine, 1-6-methylpyrazin-2-yl-4-piperidyl methylamine
PubChem CID 18525899
IUPAC Name [1-(6-methylpyrazin-2-yl)piperidin-4-yl]methanamine
SMILES CC1=CN=CC(=N1)N2CCC(CC2)CN

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 206.29
Percent Purity 97%
Chemical Name or Material [1-(6-Methylpyrazin-2-yl)piperid-4-yl]methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.