accessibility menu, dialog, popup

UserName

CH3NNNNH2

CAS RN 892502-20-4

IUPAC Name: 1-[1-(6-methylpyrazin-2-yl)piperidin-4-yl]methanamine
Synonyms: 1-[1-(6-methylpyrazin-2-yl)piperidin-4-yl]methanamine
Molecular Weight (g/mol): 206.29
Molecular Formula: C11H18N4
InChi Key: XVEZCDMRMDBGQU-UHFFFAOYSA-N
SMILES: CC1=CN=CC(=N1)N1CCC(CN)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results