missing translation for 'onlineSavingsMsg'
Learn More

1,6-Heptadien-4-ol, 97%

Product Code. 10711735 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10711735 25 g Glass bottle 25g
10096043 1 g Glass bottle 1g
10687112 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10711735 Supplier Thermo Scientific Acros Supplier No. 150090250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2883-45-6 | C7H12O | 112.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2883-45-6
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD00008663
InChI Key UTGFOWQYZKTZTN-UHFFFAOYSA-N
Synonym 1,6-heptadien-4-ol, 1,6-heptadiene-4-ol, diallylcarbinol, pubchem13740, 4-hepta-1,6-dienol, acmc-1cei5, intermediates-zcf02285, 4-hydroxy-1,6-heptadiene, 4-01-00-02249 beilstein handbook reference
PubChem CID 17902
IUPAC Name hepta-1,6-dien-4-ol
SMILES OC(CC=C)CC=C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8600g/mL
Boiling Point 151°C
Flash Point 40°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHCH2CH(OH)CH2CH=CH2
Refractive Index 1.4490 to 1.453
Quantity 25 g
Beilstein 01, 455
Specific Gravity 0.86
Formula Weight 112.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1, 6-Heptadien-4-ol, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,6-Heptadien-4-ol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.