CAS RN 2883-45-6
CAS RN 2883-45-6
IUPAC Name:
hepta-1,6-dien-4-ol
Synonyms:
hepta-1,6-dien-4-ol
Molecular Weight (g/mol):
112.17
Molecular Formula:
C7H12O
InChi Key:
UTGFOWQYZKTZTN-UHFFFAOYSA-N
SMILES:
OC(CC=C)CC=C
1
–
1
of
1
results
1
1,6-Heptadien-4-ol, 97%
CAS: 2883-45-6 Molecular Formula: C7H12O Molecular Weight (g/mol): 112.17 MDL Number: MFCD00008663 InChI Key: UTGFOWQYZKTZTN-UHFFFAOYSA-N Synonym: 1,6-heptadien-4-ol,1,6-heptadiene-4-ol,diallylcarbinol,pubchem13740,4-hepta-1,6-dienol,acmc-1cei5,intermediates-zcf02285,4-hydroxy-1,6-heptadiene,4-01-00-02249 beilstein handbook reference PubChem CID: 17902 IUPAC Name: hepta-1,6-dien-4-ol SMILES: OC(CC=C)CC=C