missing translation for 'onlineSavingsMsg'
Learn More

1,5-Pentamethylene-1H-tetrazole, 98%

Product Code. p-7058449
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372917 5 g 5g
11382917 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372917 Supplier Thermo Scientific Alfa Aesar Supplier No. L16185.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 54-95-5 | C6H10N4 | 138.174 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-95-5
Molecular Formula C6H10N4
Molecular Weight (g/mol) 138.174
MDL Number MFCD00005939
InChI Key CWRVKFFCRWGWCS-UHFFFAOYSA-N
Synonym pentylenetetrazol, pentylenetetrazole, pentetrazol, metrazole, cardiazol, cardiazole, cardiol, corazol, 1,5-pentamethylenetetrazole, corazole
PubChem CID 5917
ChEBI CHEBI:34910
IUPAC Name 6,7,8,9-tetrahydro-5H-tetrazolo[1,5-a]azepine
SMILES C1CCC2=NN=NN2CC1

Specifications

Melting Point 59°C to 61°C
Boiling Point 194°C (12 mmHg)
Quantity 5 g
UN Number UN2811
Beilstein 135492
Sensitivity Hygroscopic
Merck Index 14,7141
Formula Weight 138.17
Percent Purity 98%
Chemical Name or Material 1,5-Pentamethylene-1H-tetrazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.