missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,5-Hexadien-3-ol, 95%

Product Code. 11302056
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11302056

Brand: Alfa Aesar™ L03182.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
1,5-Hexadien-3-ol is used in the synthesis of (+)-obtusenyne. It is used as intermediates. It undergoes Sharpless kinetic resolution and ring-closing metathesis to yield nine membered oxocene.

Solubility
Sparingly Soluble.

Notes
Keep away from heat. Incompatible with Oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 924-41-4
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00008662
InChI Key SZYLTIUVWARXOO-UHFFFAOYSA-N
Synonym 1,5-hexadien-3-ol, 1,5-hexadiene-3-ol, 3-hydroxy-1,5-hexadiene
PubChem CID 79122
IUPAC Name hexa-1,5-dien-3-ol
SMILES C=CCC(C=C)O

Specifications

Density 0.877
Boiling Point 133°C to 134°C
Flash Point 29°C (84°F)
Refractive Index 1.448
Quantity 2 g
UN Number UN1987
Beilstein 1719962
Solubility Information Sparingly Soluble.
Formula Weight 98.15
Percent Purity 95%
Chemical Name or Material 1,5-Hexadien-3-ol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.