accessibility menu, dialog, popup

UserName

CAS RN 924-41-4

OHH2CCH2

CAS RN 924-41-4

IUPAC Name: hexa-1,5-dien-3-ol
Synonyms: 1,5-hexadien-3-ol
Molecular Weight (g/mol): 98.15
Molecular Formula: C6H10O
InChi Key: SZYLTIUVWARXOO-UHFFFAOYNA-N
SMILES: OC(CC=C)C=C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)