Learn More
1,3-Propanedisulfonic acid disodium salt, 99% (dry wt.)
Description
It is used as a reactant involved in synthesis of cadmium(II) bipyridine complex metal-organic frameworks with alkanedisulfonates, alkylenedisulfonate inorganic-organic hybrid materials and glycosaminoglycan mimetic involved in pharmacological studies for the treatment of amyloid A amyloidosis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is used as a reactant involved in synthesis of cadmium(II) bipyridine complex metal-organic frameworks with alkanedisulfonates, alkylenedisulfonate inorganic-organic hybrid materials and glycosaminoglycan mimetic involved in pharmacological studies for the treatment of amyloid A amyloidosis.
Solubility
Partially soluble in cold water, hot water.
Notes
Hygroscopic. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | >300°C |
| Quantity | 25 g |
| Beilstein | 3767479 |
| Sensitivity | Hygroscopic |
| Solubility Information | Partially soluble in cold water,hot water. |
| Formula Weight | 248.19 |
| Percent Purity | 99% |
| Assay | (dry wt.) |
| Chemical Name or Material | 1,3-Propanedisulfonic acid disodium salt |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.