accessibility menu, dialog, popup

UserName

NaNaOSOOSOOO

CAS RN 36589-58-9

IUPAC Name: disodium propane-1,3-disulfonate
Synonyms: disodium propane-1,3-disulfonate
Molecular Weight (g/mol): 248.17
Molecular Formula: C3H6Na2O6S2
InChi Key: DKGJFKPIUSHDIT-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]S(=O)(=O)CCCS([O-])(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)