missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Iodobenzyl)piperidine, ≥97%, Thermo Scientific™

Product Code. 10210275
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10210275 5 g 5g
10041434 250 mg 250mg
10670084 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10210275 Supplier Thermo Scientific Maybridge Supplier No. CC45211DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 859850-87-6
Molecular Formula C12H16IN
Molecular Weight (g/mol) 301.171
MDL Number MFCD06797854
InChI Key DCRZUICHKJGHQK-UHFFFAOYSA-N
Synonym 1-3-iodobenzyl piperidine, 1-3-iodophenyl methyl piperidine, piperidine,1-3-iodophenyl methyl, 1-3-iodo-benzyl-piperidine
PubChem CID 17750314
IUPAC Name 1-[(3-iodophenyl)methyl]piperidine
SMILES C1CCN(CC1)CC2=CC(=CC=C2)I

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 301.17
Percent Purity 97%
Chemical Name or Material 1-(3-Iodobenzyl)piperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.