accessibility menu, dialog, popup

UserName

IN

CAS RN 859850-87-6

IUPAC Name: 1-[(3-iodophenyl)methyl]piperidine
Synonyms: 1-[(3-iodophenyl)methyl]piperidine
Molecular Weight (g/mol): 301.17
Molecular Formula: C12H16IN
InChi Key: DCRZUICHKJGHQK-UHFFFAOYSA-N
SMILES: IC1=CC=CC(CN2CCCCC2)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results