missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethylbutyl Butanoate, TRC

Product Code. 30388721
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388721 250 mg 250mg
30384066 100 mg -
30374390 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30388721 Supplier Toronto Research Chemicals Supplier No. TRCD457700250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5332-88-7
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.26
Synonym 1,3-Dimethylbutyl ester butanoic acid, NSC 568
IUPAC Name 4-methylpentan-2-yl butanoate
SMILES CCCC(=O)OC(C)CC(C)C

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C10H20O2/c1-5-6-10(11)12-9(4)7-8(2)3/h8-9H,5-7H2,1-4H3
Formula Weight 172.1463
Chemical Name or Material 1,3-Dimethylbutyl Butanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.