CAS RN 5332-88-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,3-Dimethylbutyl Butanoate, TRC
CAS: 5332-88-7 Molecular Formula: C10H20O2 Molecular Weight (g/mol): 172.26 Synonym: 1,3-Dimethylbutyl ester butanoic acid,NSC 568 IUPAC Name: 4-methylpentan-2-yl butanoate SMILES: CCCC(=O)OC(C)CC(C)C