missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide, 98%

Product Code. 11414393
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414393 5 g 5g
11404393 1 g 1g
11424393 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11414393 Supplier Thermo Scientific Alfa Aesar Supplier No. A10962.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 22572-40-3 | C9H20N3+ | 170.28 g/mol

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide is in proteins as water soluble carboxyl modifying reagent. It is used as a crosslinking agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide is in proteins as water soluble carboxyl modifying reagent. It is used as a crosslinking agent.

Solubility
Soluble in water.

Notes
Keep in a dry, cool and well-ventilated place. Keep container tightly closed. Incompatible with acids, strong oxidizing agents, moist air and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22572-40-3
Molecular Formula C9H20N3+
Molecular Weight (g/mol) 170.28
MDL Number MFCD00011833
InChI Key JHMDMCSGQQLHNA-UHFFFAOYSA-N
Synonym 3-ethylimino methylidene amino-n,n,n-trimethylpropan-1-aminium, 1-3-trimethylaminio propyl-3-ethylcarbodiimide, 3-ethyliminomethylideneamino propyl-trimethylazanium, 1-3-dimethylaminopropyl-3-ethylcarbodiimide methiodide, 1-3-dimethylaminopropyl-3-ethylcarbodiimide, polymer-bound, 3-ethylimino methylidene amino propyl trimethylazanium, 1-3-dimethylaminopropyl-3-ethylcarbodiimide, polymer-bound, 200-400 mesh, 2 % cross-linked with divinylbenzene
PubChem CID 122243
IUPAC Name 3-(ethyliminomethylideneamino)propyl-trimethylazanium
SMILES CCN=C=NCCC[N+](C)(C)C

Specifications

Melting Point 90°C to 92°C
Quantity 5 g
Beilstein 4162760
Sensitivity Hygroscopic; Light sensitive
Solubility Information Soluble in water.
Formula Weight 297.17
Percent Purity 98%
Chemical Name or Material 1- (3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.