accessibility menu, dialog, popup

UserName

ICH3NC(?)NNCH3H3CCH3

CAS RN 22572-40-3

IUPAC Name: (3-{[(ethylimino)methylidene]amino}propyl)trimethylazanium iodide
Synonyms: (3-{[(ethylimino)methylidene]amino}propyl)trimethylazanium iodide
Molecular Weight (g/mol): 297.18
Molecular Formula: C9H20IN3
InChi Key: AGSKWMRPXWHSPF-UHFFFAOYSA-M
SMILES: [I-].CCN=C=NCCC[N+](C)(C)C
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results