missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diethoxy-1,1,3,3-tetramethyldisiloxane, TRC

Product Code. 30134862
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134862 1 g 1g
30010788 5 g 5g
30010452 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134862 Supplier Toronto Research Chemicals Supplier No. TRCD0765151G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18420-09-2
Formula Weight 222.111
InChI Formula InChI=1S/C8H22O3Si2/c1-7-9-12(3,4)11-13(5,6)10-8-2/h7-8H2,1-6H3
IUPAC Name ethoxy-[ethoxy(dimethyl)silyl]oxy-dimethylsilane
Molecular Formula C8 H22 O3 Si2
Molecular Weight (g/mol) 222.429
SMILES CCO[Si](C)(C)O[Si](C)(C)OCC
Synonym 1,3-Diethoxy-1,1,3,3-tetramethyldisiloxane, 1,1,3,3-Tetramethyl-1,3-diethoxydisiloxane, 1,3-Diethoxytetramethyldisiloxane, α,ω-Diethoxytetramethyldisiloxane;
Chemical Name or Material 1,3-Diethoxy-1,1,3,3-tetramethyldisiloxane

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.