missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diamino-2-propanol-N,N,N',N'-tetraacetic acid, 98+%, Thermo Scientific™

Product Code. 11448057
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11448057

Brand: Thermo Scientific Alfa Aesar A17463.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3148-72-9
Molecular Formula C11H18N2O9
Molecular Weight (g/mol) 322.27
MDL Number MFCD00004288
InChI Key WYMDDFRYORANCC-UHFFFAOYSA-N
Synonym dhpta, dpta, 1,3-diaminopropanol tetraacetic acid, diaminohydroxypropane tetraacetic acid, 1,3-diamino-2-propanol-n,n,n',n'-tetraacetic acid, diaminopropanol tetraacetic acid, unii-949b9zmo7m, 1,3-diamino-2-hydroxypropane-n,n,n',n'-tetraacetic acid, 2-hydroxy-1,3-propylenediaminetetraacetic acid, diaminopropanol tetra acetic acid
PubChem CID 18465
IUPAC Name 2-[[3-[bis(carboxymethyl)amino]-2-hydroxypropyl]-(carboxymethyl)amino]acetic acid
SMILES C(C(CN(CC(=O)O)CC(=O)O)O)N(CC(=O)O)CC(=O)O

Specifications

Melting Point ∼195°C (decomposition)
Quantity 5 g
Beilstein 1716800
Formula Weight 322.27
Percent Purity ≥98%
Chemical Name or Material 1,3-Diamino-2-propanol-N,N,N',N'-tetraacetic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.