accessibility menu, dialog, popup

UserName

CAS RN 3148-72-9

OHNOHOHOONOHOOHO

CAS RN 3148-72-9

IUPAC Name: 2-({3-[bis(carboxymethyl)amino]-2-hydroxypropyl}(carboxymethyl)amino)acetic acid
Synonyms: ({3-[bis(carboxymethyl)amino]-2-hydroxypropyl}(carboxymethyl)amino)acetic acid
Molecular Weight (g/mol): 322.27
Molecular Formula: C11H18N2O9
InChi Key: WYMDDFRYORANCC-UHFFFAOYSA-N
SMILES: OC(CN(CC(O)=O)CC(O)=O)CN(CC(O)=O)CC(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results