missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzothiazol-6-ylmethylamine dihydrochloride, 97%, Thermo Scientific™

Product Code. 11916791 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11916791 1 g 1g
11926791 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11916791 Supplier Thermo Scientific Acros Supplier No. 430100010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1375964-49-0
Molecular Formula C8H8N22ClH
Molecular Weight (g/mol) 237.15
InChI Key KFQZRIYKOSJQHF-UHFFFAOYSA-N
Synonym 6-benzothiazolemethanamine dihydrochloride, 1,3-benzothiazol-6-ylmethylamine dihydrochloride, 1-1,3-benzothiazol-6-yl methanamine dihydrochloride
PubChem CID 110210982
IUPAC Name 1,3-benzothiazol-6-ylmethanamine;dihydrochloride
SMILES C1=CC2=C(C=C1CN)SC=N2.Cl.Cl

Specifications

Melting Point 137.0°C to 137.5°C
Assay Percent Range 96 to max. 104% min. (Argentometry)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 237.15
Percent Purity 97%
Chemical Name or Material 1, 3-Benzothiazol-6-ylmethylamine dihydrochloride
compliance-icons
Product Identifier
  • 1,3-Benzothiazol-6-ylmethylamine dihydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.