accessibility menu, dialog, popup

UserName

CAS RN 1375964-49-0

NH2SNHClHCl

CAS RN 1375964-49-0

IUPAC Name: 1-(1,3-benzothiazol-6-yl)methanamine dihydrochloride
Synonyms: 1-(1,3-benzothiazol-6-yl)methanamine dihydrochloride
Molecular Weight (g/mol): 237.14
Molecular Formula: C8H10Cl2N2S
InChi Key: KFQZRIYKOSJQHF-UHFFFAOYSA-N
SMILES: Cl.Cl.NCC1=CC=C2N=CSC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results