missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Benzenetricarboxylic acid triallyl ester, 98%

Product Code. p-3771111 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10642825

Brand: Thermo Scientific Acros 209350010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 17832-16-5 | C18H18O6 | 330.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17832-16-5
Molecular Formula C18H18O6
Molecular Weight (g/mol) 330.33
MDL Number MFCD00008647
InChI Key VOSUIKFOFHZNED-UHFFFAOYSA-N
Synonym triallyl trimesate, triallyl benzene-1,3,5-tricarboxylate, triallyl 1,3,5-benzenetricarboxylate, triallylbenzene-1,3,5-tricarboxylate, 1,3,5-benzenetricarboxylic acid, tri-2-propenyl ester, 1,3,5-benzenetricarboxylic acid, 1,3,5-tri-2-propen-1-yl ester, 1,3,5-benzenetricarboxylic acid triallyl ester, 1,3,5-tris prop-2-en-1-yl benzene-1,3,5-tricarboxylate, triallyltrimesate, acmc-1c8hf
PubChem CID 87328
IUPAC Name tris(prop-2-enyl) benzene-1,3,5-tricarboxylate
SMILES C=CCOC(=O)C1=CC(=CC(=C1)C(=O)OCC=C)C(=O)OCC=C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 31°C to 34°C
Color Yellow
Density 1.1600g/mL
Boiling Point 212°C (2.0 mmHg)
Flash Point >113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H3(CO2CH2CH=CH2)3
Quantity 1 g
Specific Gravity 1.16
Formula Weight 330.33
Percent Purity 98%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 1, 3, 5-Benzenetricarboxylic acid triallyl ester
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.