accessibility menu, dialog, popup

UserName

CAS RN 17832-16-5

CH2OOOOH2COOCH2

CAS RN 17832-16-5

IUPAC Name: 1,3,5-tris(prop-2-en-1-yl) benzene-1,3,5-tricarboxylate
Synonyms: 1,3,5-tris(prop-2-en-1-yl) benzene-1,3,5-tricarboxylate
Molecular Weight (g/mol): 330.34
Molecular Formula: C18H18O6
InChi Key: VOSUIKFOFHZNED-UHFFFAOYSA-N
SMILES: C=CCOC(=O)C1=CC(=CC(=C1)C(=O)OCC=C)C(=O)OCC=C
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results