missing translation for 'onlineSavingsMsg'
Learn More

1-{[2-Fluoro-6-(trifluoromethyl)phenyl]methyl}-5-iodo-6-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione, TRC

Product Code. 30402464
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30402464 500 mg
30403740 100 mg
30406456 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30402464 Supplier Toronto Research Chemicals Supplier No. TRCF599485500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1150560-54-5
Molecular Formula C13H9N2O2F4I
Molecular Weight (g/mol) 428.12
Synonym Elagolix Impurity 5
SMILES CC1=C(I)C(=O)NC(=O)N1Cc2c(F)cccc2C(F)(F)F

Specifications

Quantity 500 mg
Formula Weight 427.9645
Chemical Name or Material 1-([2-Fluoro-6-(trifluoromethyl)phenyl]methyl)-5-iodo-6-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.