CAS RN 1150560-54-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-{[2-Fluoro-6-(trifluoromethyl)phenyl]methyl}-5-iodo-6-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione, TRC
CAS: 1150560-54-5 Molecular Formula: C13H9N2O2F4I Molecular Weight (g/mol): 428.12 Synonym: Elagolix Impurity 5 SMILES: CC1=C(I)C(=O)NC(=O)N1Cc2c(F)cccc2C(F)(F)F