Learn More
1,2,4-Tribromobenzene, 95%
Description
1,2,4-tribromobenzene is used in the synthesis of hyper branched poly(p-phenylene ethynylenes). It is used as cross-linking reagent during Pd-catalyzed cross-coupling of 2,5-diiodo-4-[(2-ethylhexyl)oxy]methoxybenzene and 1,4-diethynyl-2,5-bis-(octyloxy)benzene. It is a compound commonly used in organic synthesis reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,2,4-tribromobenzene is used in the synthesis of hyper branched poly(p-phenylene ethynylenes). It is used as cross-linking reagent during Pd-catalyzed cross-coupling of 2,5-diiodo-4-[(2-ethylhexyl)oxy]methoxybenzene and 1,4-diethynyl-2,5-bis-(octyloxy)benzene. It is a compound commonly used in organic synthesis reactions.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | 41°C to 43°C |
| Boiling Point | 274°C to 276°C |
| Flash Point | >110°C (230°F) |
| Quantity | 25 g |
| Beilstein | 1934823 |
| Synonym | benzene, 1,2,4-tribromo, unii-eoe1kfc260, benzene,1,2,4-tribromo, eoe1kfc260, dsstox_cid_4346, dsstox_rid_77377, dsstox_gsid_24346, 1,2,4-tribomobenzene, benzene,2,4-tribromo, acmc-1bdp2 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 314.8 |
| Percent Purity | 95% |
| Chemical Name or Material | 1,2,4-Tribromobenzene |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.