accessibility menu, dialog, popup

UserName

CAS RN 615-54-3

BrBrBr

CAS RN 615-54-3

IUPAC Name: 1,2,4-tribromobenzene
Synonyms: 1,2,4-tribromobenzene
Molecular Weight (g/mol): 314.8
Molecular Formula: C6H3Br3
InChi Key: FWAJPSIPOULHHH-UHFFFAOYSA-N
SMILES: BrC1=CC=C(Br)C(Br)=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results