missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Tribromobenzene, 95%

Product Code. p-7027205
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11470228

Brand: Thermo Scientific Alfa Aesar B23778.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 615-54-3 | C6H3Br3 | 314.802 g/mol

1,2,4-tribromobenzene is used in the synthesis of hyper branched poly(p-phenylene ethynylenes). It is used as cross-linking reagent during Pd-catalyzed cross-coupling of 2,5-diiodo-4-[(2-ethylhexyl)oxy]methoxybenzene and 1,4-diethynyl-2,5-bis-(octyloxy)benzene. It is a compound commonly used in organic synthesis reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,4-tribromobenzene is used in the synthesis of hyper branched poly(p-phenylene ethynylenes). It is used as cross-linking reagent during Pd-catalyzed cross-coupling of 2,5-diiodo-4-[(2-ethylhexyl)oxy]methoxybenzene and 1,4-diethynyl-2,5-bis-(octyloxy)benzene. It is a compound commonly used in organic synthesis reactions.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-54-3
Molecular Formula C6H3Br3
Molecular Weight (g/mol) 314.802
MDL Number MFCD00000062
InChI Key FWAJPSIPOULHHH-UHFFFAOYSA-N
PubChem CID 12002
IUPAC Name 1,2,4-tribromobenzene
SMILES C1=CC(=C(C=C1Br)Br)Br

Specifications

Melting Point 41°C to 43°C
Boiling Point 274°C to 276°C
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 1934823
Synonym benzene, 1,2,4-tribromo, unii-eoe1kfc260, benzene,1,2,4-tribromo, eoe1kfc260, dsstox_cid_4346, dsstox_rid_77377, dsstox_gsid_24346, 1,2,4-tribomobenzene, benzene,2,4-tribromo, acmc-1bdp2
Solubility Information Insoluble in water.
Formula Weight 314.8
Percent Purity 95%
Chemical Name or Material 1,2,4-Tribromobenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.