missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Tris(2-cyanoethoxy)propane, 97%, Thermo Scientific™

Product Code. 11354227
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11354227

Brand: Thermo Scientific Alfa Aesar 036702.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2465-93-2
Molecular Formula C12H17N3O3
Molecular Weight (g/mol) 251.286
MDL Number MFCD00019868
InChI Key ALGVJKNIAOBBBJ-UHFFFAOYSA-N
Synonym 1,2,3-tris 2-cyanoethoxy propane, propanenitrile, 3,3',3-1,2,3-propanetriyltris oxy tris, 3,3',3-propane-1,2,3-triyltrioxytripropiononitrile, tris .beta.-cyanoethoxy propane, glycerol tris 2-cyanoethyl ether, propionitrile, 3,3',3-1,2,3-propanetriyltrioxy tri, 3-2,3-bis 2-cyanoethoxy propoxy propanenitrile, 3-1,3-bis 2-cyanoethoxy propan-2-yl oxy propanenitrile, propanenitrile,3,3',3-1,2,3-propanetriyltris oxy tris
PubChem CID 75572
IUPAC Name 3-[2,3-bis(2-cyanoethoxy)propoxy]propanenitrile
SMILES C(COCC(COCCC#N)OCCC#N)C#N

Specifications

Density 1.107 g/mL
Quantity 25 g
UN Number UN3276
Solubility Information Soluble in water.
Formula Weight 251.29
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1,2,3-Tris(2-cyanoethoxy)propane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.