CAS RN 2465-93-2
IUPAC Name:
3-{[1,3-bis(2-cyanoethoxy)propan-2-yl]oxy}propanenitrile
Synonyms:
3-{[1,3-bis(2-cyanoethoxy)propan-2-yl]oxy}propanenitrile
Molecular Weight (g/mol):
251.29
Molecular Formula:
C12H17N3O3
InChi Key:
ALGVJKNIAOBBBJ-UHFFFAOYSA-N
SMILES:
N#CCCOCC(COCCC#N)OCCC#N
1
–
1
of
1
results
1
Alfa Aesar™ 1,2,3-Tris(2-cyanoethoxy)propane, 97%, Thermo Scientific™
CAS: 2465-93-2 Molecular Formula: C12H17N3O3 Molecular Weight (g/mol): 251.286 MDL Number: MFCD00019868 InChI Key: ALGVJKNIAOBBBJ-UHFFFAOYSA-N Synonym: 1,2,3-tris 2-cyanoethoxy propane,propanenitrile, 3,3',3-1,2,3-propanetriyltris oxy tris,3,3',3-propane-1,2,3-triyltrioxytripropiononitrile,tris .beta.-cyanoethoxy propane,glycerol tris 2-cyanoethyl ether,propionitrile, 3,3',3-1,2,3-propanetriyltrioxy tri,3-2,3-bis 2-cyanoethoxy propoxy propanenitrile,3-1,3-bis 2-cyanoethoxy propan-2-yl oxy propanenitrile,propanenitrile,3,3',3-1,2,3-propanetriyltris oxy tris PubChem CID: 75572 IUPAC Name: 3-[2,3-bis(2-cyanoethoxy)propoxy]propanenitrile SMILES: C(COCC(COCCC#N)OCCC#N)C#N