missing translation for 'onlineSavingsMsg'
Learn More

1-(2,3-Dihydro-1H-indol-1-yl)prop-2-en-1-one, TRC

Product Code. 30387737
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387737 50 mg -
30379444 100 mg 100mg
30382876 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387737 Supplier Toronto Research Chemicals Supplier No. TRCD45301350MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104902-94-5
Molecular Formula C11H11NO
Molecular Weight (g/mol) 173.21
SMILES C=CC(=O)N1CCc2ccccc12

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C11H11NO/c1-2-11(13)12-8-7-9-5-3-4-6-10(9)12/h2-6H,1,7-8H2
Formula Weight 173.0841
Chemical Name or Material 1-(2,3-Dihydro-1H-indol-1-yl)prop-2-en-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.