missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetra-O-acetyl-beta-D-ribopyranose, 98%, Thermo Scientific™

Product Code. 15408059
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15408059 250 mg 250mg
15498049 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15408059 Supplier Thermo Scientific Alfa Aesar Supplier No. J66623.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4049-34-7
Molecular Formula C13H18O9
Molecular Weight (g/mol) 318.278
MDL Number MFCD00006596
InChI Key MJOQJPYNENPSSS-LPWJVIDDSA-N
Synonym tetra-o-acetyl-beta-d-ribopyranose, 2s,3r,4r,5r-tetrahydro-2h-pyran-2,3,4,5-tetrayl tetraacetate, 1,2,3,4-tetra-o-acetyl-b-d-ribopyranose, beta-d-ribopyranose 1,2,3,4-tetraacetate, 3r,4r,5r,6s-4,5,6-triacetyloxyoxan-3-yl acetate, 2s,3r,4r,5r-2,4,5-tris acetyloxy oxan-3-yl acetate, beta-d-ribopyranose tetraacetate, mjoqjpynenpsss-lpwjviddsa, 1,2,3,4-tetra-o-acetyl-, a-d-ribopyranose, b-d-ribopyranose 1,2,3,4-tetraacetate
PubChem CID 97336
IUPAC Name [(3R,4R,5R,6S)-4,5,6-triacetyloxyoxan-3-yl] acetate
SMILES CC(=O)OC1COC(C(C1OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point 110°C to 112°C
Color White
Quantity 250 mg
Optical Rotation +55° (c=3 in Methanol)
Formula Weight 318.28
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1,2,3,4-Tetra-O-acetyl-beta-D-ribopyranose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.