accessibility menu, dialog, popup

UserName

CAS RN 4049-34-7

IUPAC Name: 3,4,5-tris(acetyloxy)oxan-2-yl acetate
Synonyms: 3,4,5-tris(acetyloxy)oxan-2-yl acetate
Molecular Weight (g/mol): 318.28
Molecular Formula: C13H18O9
InChi Key: MJOQJPYNENPSSS-UHFFFAOYNA-N
SMILES: CC(=O)OC1COC(OC(C)=O)C(OC(C)=O)C1OC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results