missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4,7,8-Hexachlorodibenzodioxin (>90%), TRC

Product Code. 30407214
Change view
Click to view available options
Quantity:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30407214 250 mg
30402537 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30407214 Supplier Toronto Research Chemicals Supplier No. TRCH290780250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39227-28-6
Molecular Formula C12 H2 Cl6 O2
Molecular Weight (g/mol) 390.86
Synonym Dibenzo[b,e][1,4]dioxin, 1,2,3,4,7,8-hexachloro-, 1,2,3,4,7,8-Hexachlorodibenzo[b,e][1,4]dioxin, 1,2,3,4,7,8-Hexachlorodibenzo-p-dioxin, 1,2,3,4,7,8-Hexachlorodibenzo[1,4]dioxin, 1,2,3,4,7,8-Hexachlorodibenzodioxin, 1,2,3,4,7,8-HxCDD, 123478-HxCDD, D 66, PCDD 66
IUPAC Name 1,2,3,4,7,8-hexachlorodibenzo-p-dioxin
SMILES Clc1cc2Oc3c(Cl)c(Cl)c(Cl)c(Cl)c3Oc2cc1Cl

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C12H2Cl6O2/c13-3-1-5-6(2-4(3)14)20-12-10(18)8(16)7(15)9(17)11(12)19-5/h1-2H
Formula Weight 387.8186
Chemical Name or Material 1,2,3,4,7,8-Hexachlorodibenzo-p-dioxin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.