CAS RN 39227-28-6
IUPAC Name:
1,2,3,4,7,8-hexachlorooxanthrene
Synonyms:
1,2,3,4,7,8-hexachlorooxanthrene
Molecular Weight (g/mol):
390.85
Molecular Formula:
C12H2Cl6O2
InChi Key:
WCYYQNSQJHPVMG-UHFFFAOYSA-N
SMILES:
ClC1=CC2=C(OC3=C(Cl)C(Cl)=C(Cl)C(Cl)=C3O2)C=C1Cl
1
–
1
of
1
results
1
1,2,3,4,7,8-Hexachlorodibenzodioxin (>90%), TRC
CAS: 39227-28-6 Molecular Formula: C12 H2 Cl6 O2 Molecular Weight (g/mol): 390.86 Synonym: Dibenzo[b,e][1,4]dioxin, 1,2,3,4,7,8-hexachloro-,1,2,3,4,7,8-Hexachlorodibenzo[b,e][1,4]dioxin,1,2,3,4,7,8-Hexachlorodibenzo-p-dioxin,1,2,3,4,7,8-Hexachlorodibenzo[1,4]dioxin,1,2,3,4,7,8-Hexachlorodibenzodioxin,1,2,3,4,7,8-HxCDD,123478-HxCDD,D 66,PCDD 66 IUPAC Name: 1,2,3,4,7,8-hexachlorodibenzo-p-dioxin SMILES: Clc1cc2Oc3c(Cl)c(Cl)c(Cl)c(Cl)c3Oc2cc1Cl