missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4,7,7-Hexachloro-5-phenyl-2-norbornene, TRC

Product Code. 30396301
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396301 50 mg 50mg
30405117 100 mg 100mg
30391182 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30396301 Supplier Toronto Research Chemicals Supplier No. TRCH29082050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17064-54-9
Molecular Formula C13H8Cl6
Molecular Weight (g/mol) 376.92
Synonym 1,2,3,4,7,7-Hexachloro-5-phenyl-bicyclo[2.2.1]hept-2-ene
IUPAC Name (1R,4S)-1,2,3,4,7,7-hexachloro-5-phenylbicyclo[2.2.1]hept-2-ene
SMILES ClC1=C(Cl)[C@@]2(Cl)C(C[C@]1(Cl)C2(Cl)Cl)c3ccccc3

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C13H8Cl6/c14-9-10(15)12(17)8(7-4-2-1-3-5-7)6-11(9,16)13(12,18)19/h1-5,8H,6H2/t8?,11-,12+/m1/s1
Formula Weight 373.8757
Chemical Name or Material 1,2,3,4,7,7-Hexachloro-5-phenyl-2-norbornene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.