CAS RN 17064-54-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,2,3,4,7,7-Hexachloro-5-phenyl-2-norbornene, TRC
CAS: 17064-54-9 Molecular Formula: C13H8Cl6 Molecular Weight (g/mol): 376.92 Synonym: 1,2,3,4,7,7-Hexachloro-5-phenyl-bicyclo[2.2.1]hept-2-ene IUPAC Name: (1R,4S)-1,2,3,4,7,7-hexachloro-5-phenylbicyclo[2.2.1]hept-2-ene SMILES: ClC1=C(Cl)[C@@]2(Cl)C(C[C@]1(Cl)C2(Cl)Cl)c3ccccc3