missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Bis(diphenylphosphino)ferrocene, 97%, Thermo Scientific Chemicals

Product Code. 11454657
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454657 1 g 1g
11464657 5 g 5g
11474657 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11454657 Supplier Thermo Scientific Alfa Aesar Supplier No. B21166.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

1,1'-Bis(diphenylphosphino)ferrocene acts as a ligand in homogeneous catalysis. It is used as a ligand for ruthenium-catalyzed greener amine synthesis from amines and alcohols by hydrogen-borrowing. It is also employed as a ligand for Buchwald-Hartwig cross coupling. Further, it is used in the synthesis of coordination compound as well as the formation of complexes with various metals such as palladium chloride. In addition to this, it serves as a reagent for palladium-catalyzed coupling reactions and also plays an important role in Suzuki reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1′-Bis(diphenylphosphino)ferrocene acts as a ligand in homogeneous catalysis. It is used as a ligand for ruthenium-catalyzed greener amine synthesis from amines and alcohols by hydrogen-borrowing. It is also employed as a ligand for Buchwald-Hartwig cross coupling. Further, it is used in the synthesis of coordination compound as well as the formation of complexes with various metals such as palladium chloride. In addition to this, it serves as a reagent for palladium-catalyzed coupling reactions and also plays an important role in Suzuki reaction.

Solubility
Soluble in chloroform, dichloromethane, alcohol and pentane. Insoluble in water.

Notes
Air sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12150-46-8
Molecular Formula C34H28FeP2
Molecular Weight (g/mol) 554.39
MDL Number MFCD00001422
InChI Key KZPYGQFFRCFCPP-UHFFFAOYSA-N
IUPAC Name λ²-iron(2+) bis(1-(diphenylphosphanyl)cyclopenta-2,4-dien-1-ide)
SMILES [Fe].c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∽182°C (decomposition)
Assay Percent Range 0.97
Quantity 1 g
Sensitivity Air sensitive
Solubility Information Soluble in chloroform,dichloromethane,alcohol and pentane. Insoluble in water.
Formula Weight 554.4
Percent Purity 97%
Chemical Name or Material 1, 1'-Bis(diphenylphosphino)ferrocene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.