accessibility menu, dialog, popup

UserName

FePP

CAS RN 12150-46-8

IUPAC Name: λ²-iron(2+) bis(1-(diphenylphosphanyl)cyclopenta-2,4-dien-1-ide)
Synonyms: λ²-iron(2+) bis(1-(diphenylphosphanyl)cyclopenta-2,4-dien-1-ide)
Molecular Weight (g/mol): 554.39
Molecular Formula: C34H28FeP2
InChi Key: KZPYGQFFRCFCPP-UHFFFAOYSA-N
SMILES: [Fe++].C1=C[C-](C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=C[C-](C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)