missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Patched 1/PTCH Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NBP1-71662
This item is not returnable.
View return policy
Description
Patched 1/PTCH Polyclonal specifically detects Patched 1/PTCH in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Patched 1/PTCH | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| BCNSFLJ42602, FLJ26746, HPE7, NBCCS, patched (Drosophila) homolog, patched 1, patched homolog (Drosophila), patched homolog 1, patched homolog 1 (Drosophila), protein patched homolog 1, PTC, PTC1, PTCH, PTCH protein +12b, PTCH protein +4', PTCH protein -10, PTCH11 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.22 mg/mL | |
| Western Blot 1:2500, Immunohistochemistry 1:50 - 1:250, Immunohistochemistry-Paraffin 1:50 - 1:250 | |
| Q13635 | |
| PTCH1 | |
| A genomic peptide made to a C-terminal region of the human Patched 1 protein (within residues 1200-1350). [Swiss-Prot Q13635] | |
| 0.1 mL | |
| Cell Cycle and Replication, Stem Cell Signaling Pathway | |
| 5727 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction