missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Patched 1/PTCH Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
£159.00 - £367.00
Specifications
| Antigen | Patched 1/PTCH |
|---|---|
| Concentration | 1.22 mg/mL |
| Dilution | Western Blot 1:2500, Immunohistochemistry 1:50 - 1:250, Immunohistochemistry-Paraffin 1:50 - 1:250 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18213414
|
Novus Biologicals
NBP1-71662SS |
0.025 mL |
£159.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18776083
|
Novus Biologicals
NBP1-71662 |
0.1 mL |
£367.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Patched 1/PTCH Polyclonal specifically detects Patched 1/PTCH in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Patched 1/PTCH | |
| Western Blot 1:2500, Immunohistochemistry 1:50 - 1:250, Immunohistochemistry-Paraffin 1:50 - 1:250 | |
| Polyclonal | |
| Rabbit | |
| Cell Cycle and Replication, Stem Cell Signaling Pathway | |
| PBS with 0.05% Sodium Azide | |
| BCNSFLJ42602, FLJ26746, HPE7, NBCCS, patched (Drosophila) homolog, patched 1, patched homolog (Drosophila), patched homolog 1, patched homolog 1 (Drosophila), protein patched homolog 1, PTC, PTC1, PTCH, PTCH protein +12b, PTCH protein +4', PTCH protein -10, PTCH11 | |
| PTCH1 | |
| IgG | |
| Affinity Purified |
| 1.22 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| Q13635 | |
| 5727 | |
| A genomic peptide made to a C-terminal region of the human Patched 1 protein (within residues 1200-1350). [Swiss-Prot Q13635] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title