Learn More
OPA1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 27 publications
Brand:  Novus Biologicals NB110-55290
 View more versions of this product
Product Code. 18744993
  • £441.00 / 0.10mL
Description
OPA1 Polyclonal specifically detects OPA1 in Human, Mouse, Rat, Porcine, Chicken, Zebrafish samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoblotting.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
OPA1
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
FLJ12460, KIAA0567dynamin-like 120 kDa protein, mitochondrial, MGM1, mitochondrial dynamin-like GTPase, NPGlargeG, NTGmitochondrial dynamin-like 120 kDa protein, optic atrophy 1 (autosomal dominant), Optic atrophy protein 1
Rabbit
111 kDa
0.1 mL
Apoptosis, Mitochondrial Fusion Proteins, Neurodegeneration, Neuroscience, Sensory Systems, Vision
4976
Human, Mouse, Rat, Pig, Chicken
IgG
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot
1 mg/mL
Western Blot 2 ug/ml, Simple Western 20 ug/ml, Immunohistochemistry 2.5 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 34923139), Immunohistochemistry-Paraffin 2.5 ug/ml, Immunoblotting reported in scientific literature (PMID 25224038)
O60313
OPA1
A synthetic peptide made to an internal region within residues 500-600 of human OPA1. [Swiss-Prot: O60313]
Affinity Purified
RUO
Primary
Human, mouse, rat and chicken. Predicted to react with primate based on 100% sequence homology. Immunogen sequence has 94% homology to Zebrafish.
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.