missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OPA1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 27 publications
£184.00 - £441.00
Specifications
| Antigen | OPA1 |
|---|---|
| Dilution | Western Blot 2 ug/ml, Simple Western 20 ug/ml, Immunohistochemistry 2.5 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 34923139), Immunohistochemistry-Paraffin 2.5 ug/ml, Immunoblotting reported in scientific literature (PMID 25224038) |
| Applications | Immunocytochemistry |
| Target Species | Pig |
| Gene Alias | FLJ12460, KIAA0567dynamin-like 120 kDa protein, mitochondrial, MGM1, mitochondrial dynamin-like GTPase, NPGlargeG, NTGmitochondrial dynamin-like 120 kDa protein, optic atrophy 1 (autosomal dominant), Optic atrophy protein 1 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18746373
|
Novus Biologicals
NB110-55290SS |
£184.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18744993
|
Novus Biologicals
NB110-55290 |
0.1 mL |
£441.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
OPA1 Polyclonal specifically detects OPA1 in Human, Mouse, Rat, Porcine, Chicken, Zebrafish samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoblotting.Specifications
| OPA1 | |
| Immunocytochemistry | |
| FLJ12460, KIAA0567dynamin-like 120 kDa protein, mitochondrial, MGM1, mitochondrial dynamin-like GTPase, NPGlargeG, NTGmitochondrial dynamin-like 120 kDa protein, optic atrophy 1 (autosomal dominant), Optic atrophy protein 1 | |
| IgG | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot 2 ug/ml, Simple Western 20 ug/ml, Immunohistochemistry 2.5 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 34923139), Immunohistochemistry-Paraffin 2.5 ug/ml, Immunoblotting reported in scientific literature (PMID 25224038) | |
| Pig | |
| OPA1 | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title