missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NK1R Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 35 publications
Brand: Novus Biologicals NB300-119
This item is not returnable.
View return policy
Description
NK1R Polyclonal specifically detects NK1R in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| NK1R | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| NK-1R, NK1Rneurokinin 1 receptor, NKIR, SPRsubstance-P receptor, TAC1Rtachykinin 1 receptor (substance P receptor, neurokinin 1 receptor), tachykinin receptor 1 (substance P receptor; neurokinin-1 receptor), tachykinin receptor 1NK-1 receptor | |
| Rabbit | |
| 46 kDa | |
| 0.1 mL | |
| GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 6869 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1-2 ug/ml, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 10 ug/ml, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 10 ug/ml, Immunohistochemistry-Frozen reported in scientific literature (PMID 17604979) | |
| P25103 | |
| TACR1 | |
| A peptide derived from the N-terminus region between amino acids 1-50 of human Neurokinin 1 Receptor. [UniProt# P25103] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Reacts with human, mouse and rat. Immunogen has 94% sequence identity with guinea pig. Mouse reactivity is from a customer review. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction