missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NK1R Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 35 publications
£242.00 - £500.00
Specifications
| Antigen | NK1R |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 10 ug/ml, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 10 ug/ml, Immunohistochemistry-Frozen reported in scientific literature (PMID 17604979) |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18275450
|
Novus Biologicals
NB300-119SS |
0.025 mL |
£242.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18779273
|
Novus Biologicals
NB300-119 |
0.1 mL |
£500.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NK1R Polyclonal specifically detects NK1R in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| NK1R | |
| Western Blot 1-2 ug/ml, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 10 ug/ml, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 10 ug/ml, Immunohistochemistry-Frozen reported in scientific literature (PMID 17604979) | |
| Polyclonal | |
| Rabbit | |
| GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| PBS with 0.02% Sodium Azide | |
| NK-1R, NK1Rneurokinin 1 receptor, NKIR, SPRsubstance-P receptor, TAC1Rtachykinin 1 receptor (substance P receptor, neurokinin 1 receptor), tachykinin receptor 1 (substance P receptor; neurokinin-1 receptor), tachykinin receptor 1NK-1 receptor | |
| TACR1 | |
| IgG | |
| Affinity Purified | |
| Reacts with human, mouse and rat. Immunogen has 94% sequence identity with guinea pig. Mouse reactivity is from a customer review. |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| P25103 | |
| 6869 | |
| A peptide derived from the N-terminus region between amino acids 1-50 of human Neurokinin 1 Receptor. [UniProt# P25103] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 46 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title