missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GATA-4 Antibody (6H10), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-47540
This item is not returnable.
View return policy
Description
GATA-4 Monoclonal specifically detects GATA-4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| GATA-4 | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.03% Sodium Azide | |
| GATA binding protein 4, GATA-binding factor 4, GATA-binding protein 4, MGC126629, transcription factor GATA-4 | |
| Mouse | |
| 46 kDa | |
| 0.1 mL | |
| Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| 2626 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
| Western Blot, ELISA | |
| 6H10 | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| P43694 | |
| GATA4 | |
| Purified recombinant fragment of human GATA4 expressed in E. coli. [UniProt# P43694] | |
| Unpurified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction