missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GATA-4 Antibody (6H10), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
£165.00 - £367.00
Specifications
| Antigen | GATA-4 |
|---|---|
| Clone | 6H10 |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18282173
|
Novus Biologicals
NBP1-47540SS |
0.025 mL |
£165.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18740144
|
Novus Biologicals
NBP1-47540 |
0.1 mL |
£367.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GATA-4 Monoclonal specifically detects GATA-4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
| GATA-4 | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human, Mouse, Rat | |
| P43694 | |
| 2626 | |
| Purified recombinant fragment of human GATA4 expressed in E. coli. [UniProt# P43694] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 6H10 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| Ascites with 0.03% Sodium Azide | |
| GATA binding protein 4, GATA-binding factor 4, GATA-binding protein 4, MGC126629, transcription factor GATA-4 | |
| GATA4 | |
| IgG2b | |
| Unpurified | |
| 46 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title