missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BNIP3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-77683
This item is not returnable.
View return policy
Description
BNIP3 Polyclonal specifically detects BNIP3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| BNIP3 | |
| Polyclonal | |
| Unconjugated | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| BCL2/adenovirus E1B 19 kDa protein-interacting protein 3, BCL2/adenovirus E1B 19kDa interacting protein 3, BCL2/adenovirus E1B 19kD-interacting protein 3, NIP3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.17 mg/mL | |
| Western Blot reported in scientific literature (PMID 19574421), Flow Cytometry 2-5 ug/million cells/ml, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100 | |
| Q12983 | |
| BNIP3 | |
| A synthetic peptide made to an internal region of the human BNIP3 protein (within residues 50-150). [Swiss-Prot Q12983] | |
| 0.1 mL | |
| Apoptosis, Autophagy, Cancer, Hypoxia, Tumor Suppressors | |
| 664 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction