missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BNIP3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
£169.00 - £395.00
Specifications
| Antigen | BNIP3 |
|---|---|
| Concentration | 1.17 mg/mL |
| Dilution | Western Blot reported in scientific literature (PMID 19574421), Flow Cytometry 2-5 ug/million cells/ml, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18247145
|
Novus Biologicals
NBP1-77683SS |
0.025 mL |
£169.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18759153
|
Novus Biologicals
NBP1-77683 |
0.1 mL |
£395.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BNIP3 Polyclonal specifically detects BNIP3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| BNIP3 | |
| Western Blot reported in scientific literature (PMID 19574421), Flow Cytometry 2-5 ug/million cells/ml, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100 | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Autophagy, Cancer, Hypoxia, Tumor Suppressors | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| BCL2/adenovirus E1B 19 kDa protein-interacting protein 3, BCL2/adenovirus E1B 19kDa interacting protein 3, BCL2/adenovirus E1B 19kD-interacting protein 3, NIP3 | |
| BNIP3 | |
| IgG | |
| Affinity Purified |
| 1.17 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| Q12983 | |
| 664 | |
| A synthetic peptide made to an internal region of the human BNIP3 protein (within residues 50-150). [Swiss-Prot Q12983] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title